siRNA Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178874.2 | NP_849205.1 | 2, 3 | 838 | ||
| GenBank mRNA | AK157378.1 | - | 2, 3 | 951 | ||
| AK172957.1 | - | 2, 3 | 693 | |||
| BC052035.1 | - | 2, 3 | 838 | 5695005 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmcc2| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178874.2 | NP_849205.1 | 2, 3 | 838 | ||
| GenBank mRNA | AK157378.1 | - | 2, 3 | 951 | ||
| AK172957.1 | - | 2, 3 | 693 | |||
| BC052035.1 | - | 2, 3 | 838 | 5695005 | ||