siRNA Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178892.5 | NP_849223.2 | 2 | 707 | ||
| GenBank mRNA | AK028529.1 | - | 2 | 708 | ||
| AK032453.1 | - | 2 | 691 | |||
| AK150248.1 | - | 2 | 689 | |||
| AK162094.1 | - | 2 | 713 | |||
| AK163347.1 | - | 2 | 747 | |||
| BC068173.1 | - | 2 | 681 | 30616032 | ||

Tiparp
Genomic Map