siRNA Details
Gene Symbol: AA792892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178894.4 | NP_849225.2 | 3 | 532 | ||
GenBank mRNA | AK053948.1 | - | 487 | |||
AK087752.1 | - | 556 | ||||
BC120516.1 | - | 3 | 532 | 40129439 | ||
BC125434.1 | - | 3 | 532 | 40129949 |
Gene Symbol: C87414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164284.1 | NP_001157756.1 | 6 | 1287 | ||
NM_001164285.1 | NP_001157757.1 | 4 | 1140 | |||
GenBank mRNA | AK136012.1 | - | 4 | 1097 | ||
AK139835.1 | - | 6 | 1337 | |||
AK163210.1 | - | 6 | 779 | |||
AK163343.1 | - | 1220 | ||||
BC052888.1 | - | 5 | 706 | 30051439 | ||
BC080704.1 | - | 5 | 1245 | 30476802 | ||
BC083101.1 | - | 4 | 1096 | 30477931 | ||
BC085232.1 | - | 6 | 1220 | 30476183 |
Gene Symbol: Gm3259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270456.1 | NP_001257385.1 | 5 | 1221 | ||
GenBank mRNA | DV656723.1 | - | 1 | 18 |
Gene Symbol: Gm6367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054302.1 | - | 1122 |
