siRNA Details
| Gene Symbol: Zfp324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178732.3 | NP_848847.3 | 4 | 981 | ||
| GenBank mRNA | AK044240.1 | - | 4 | 978 | ||
| AK051488.1 | - | 4 | 1004 | |||
| AK052251.1 | - | 3 | 874 | |||
| AK054407.1 | - | 4 | 1026 | |||
| AK085053.1 | - | 3 | 876 | |||
| AK152107.1 | - | 4 | 1007 | |||
| AK152696.1 | - | 4 | 924 | |||
| AK165922.1 | - | 4 | 977 | |||
| BC053093.1 | - | 4 | 981 | 6400158 | ||

Zfp324
Genomic Map