siRNA Details
| Gene Symbol: Zfp750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178763.4 | NP_848878.1 | 2 | 633 | ||
| GenBank mRNA | AK028879.1 | - | 2 | 582 | ||
| AK030690.1 | - | 2 | 588 | |||
| AK037185.1 | - | 2 | 594 | |||
| AK037240.1 | - | 2 | 579 | |||
| AK076268.1 | - | 2 | 625 | |||
| AK140792.1 | - | 625 | ||||
| BC080834.1 | - | 2 | 604 | |||
| BC150905.1 | - | 1 | 487 | 9087816 | ||

Zfp750
Genomic Map