siRNA Details
| Gene Symbol: Tmem72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178768.4 | NP_848883.2 | 3 | 358 | ||
| GenBank mRNA | AK033856.1 | - | 3 | 494 | ||
| AK049048.1 | - | 3 | 513 | |||
| AK049938.1 | - | 3 | 471 | |||
| AK085603.1 | - | 3 | 359 | |||
| AK141578.1 | - | 3 | 515 | |||
| BC093486.1 | - | 3 | 501 | 30697044 | ||
| BC116440.1 | - | 3 | 342 | 40104615 | ||
| BC116441.1 | - | 3 | 342 | 40104616 | ||

Tmem72
Genomic Map