siRNA Details
| Gene Symbol: Iffo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028273.1 | - | 2 | 808 | ||
| AK031129.1 | - | 2 | 788 | |||
| AK044709.1 | - | 2 | 805 | |||
| AK044728.1 | - | 2 | 798 | |||
| AK053568.1 | - | 2 | 805 | |||
| AK163571.1 | - | 2 | 807 | |||
| AK169554.1 | - | 1 | 32 | |||
| BC057601.1 | - | 2 | 247 | 6403474 | ||

Iffo1
Genomic Map