siRNA Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170855.1 | NP_001164326.1 | 2, 3 | 548 | ||
| NM_178872.4 | NP_849203.2 | 2, 3 | 549 | |||
| GenBank mRNA | AB103063.1 | - | 2, 3 | 343 | ||
| AY251389.1 | - | 2, 3 | 306 | |||
| BC058106.1 | - | 2, 3 | 467 | 6836197 | ||
| CJ046908.1 | - | 2, 3 | 547 | |||

Trim36
Genomic Map