siRNA Details
| Gene Symbol: Dcun1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190733.1 | NP_001177662.1 | 4, 5 | 391 | ||
| NM_001190734.1 | NP_001177663.1 | 5, 6 | 326 | |||
| NM_178896.5 | NP_849227.1 | 4, 5 | 391 | |||
| GenBank mRNA | AK033559.1 | - | 4, 5 | 336 | ||
| AK076991.1 | - | 5, 6 | 327 | |||
| BC060213.1 | - | 4, 5 | 333 | |||
| BC108346.1 | - | 4, 5 | 293 | 6310392 | ||
| BC137627.1 | - | 4, 5 | 335 | 8860646 | ||
| BC137628.1 | - | 4, 5 | 335 | 8860647 | ||
| BC144794.1 | - | 4, 5 | 335 | 9053342 | ||

Dcun1d4
Genomic Map