siRNA Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113399.1 | NP_001106870.1 | 6 | 996 | ||
| NM_001113400.1 | NP_001106871.1 | 6 | 1018 | |||
| NM_178723.6 | NP_848838.2 | 8 | 1528 | |||
| GenBank mRNA | AK046798.1 | - | 8 | 1529 | ||
| AK050293.1 | - | 6 | 1019 | |||
| AK078551.1 | - | 6 | 973 | |||
| AK079322.1 | - | 8 | 1413 | |||
| AK149352.1 | - | 8 | 1509 | |||
| BC132352.1 | - | 6 | 1107 | 40130629 | ||
| BC138239.1 | - | 6 | 1107 | 8861258 | ||

Zfp385b
Genomic Map