siRNA Details
| Gene Symbol: Klhl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178741.3 | NP_848856.1 | 10 | 3051 | ||
| GenBank mRNA | AK003977.1 | - | 1 | 914 | ||
| AK031028.1 | - | 10 | 3052 | |||
| AK034618.1 | - | 10 | 3016 | |||
| AK049960.1 | - | 9 | 2939 | |||
| BC086802.1 | - | 9 | 2912 | 30612176 | ||

Klhl8
Genomic Map