siRNA Details
| Gene Symbol: Lppr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178756.4 | NP_848871.1 | 8 | 2215 | ||
| GenBank mRNA | AK087439.1 | - | 8 | 2366 | ||
| AK087514.1 | - | 8 | 2216 | |||
| AK160897.1 | - | 8 | 2342 | |||
| AY345342.1 | - | 8 | 2215 | |||
| BC096762.1 | - | 8 | 2191 | 30094664 | ||

Lppr1
Genomic Map