siRNA Details
| Gene Symbol: Zfp750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178763.4 | NP_848878.1 | 3 | 2864 | ||
| GenBank mRNA | AK030690.1 | - | 3 | 2819 | ||
| AK037185.1 | - | 3 | 2825 | |||
| AK037240.1 | - | 3 | 2811 | |||
| AK140792.1 | - | 2859 | ||||

Zfp750
Genomic Map