siRNA Details
| Gene Symbol: Slc25a40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044260.1 | - | 1432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a40| Gene Symbol: Slc25a40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044260.1 | - | 1432 | |||