siRNA Details
| Gene Symbol: Tmem72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178768.4 | NP_848883.2 | 5 | 1192 | ||
| GenBank mRNA | AK033856.1 | - | 5 | 1329 | ||
| AK049048.1 | - | 5 | 1347 | |||
| AK049938.1 | - | 5 | 1305 | |||
| AK085603.1 | - | 5 | 1193 | |||
| AK141578.1 | - | 5 | 1349 | |||
| BC093486.1 | - | 5 | 1335 | 30697044 | ||
| BC116440.1 | - | 5 | 1176 | 40104615 | ||
| BC116441.1 | - | 5 | 1176 | 40104616 | ||

Tmem72
Genomic Map