siRNA Details
| Gene Symbol: Skint4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178786.4 | NP_848901.2 | 3 | 585 | ||
| GenBank mRNA | AK028550.1 | - | 3 | 636 | ||
| AK028795.1 | - | 3 | 586 | |||
| AK035748.1 | - | 3 | 503 | |||
| AK162813.1 | - | 4 | 600 | |||
| EF494897.1 | - | 3 | 384 | |||
| EF494898.1 | - | 3 | 384 | |||

Skint4
Genomic Map