siRNA Details
| Gene Symbol: Tmem241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289666.1 | NP_001276595.1 | 15 | 2215 | ||
| NM_001289667.1 | NP_001276596.1 | 13 | 2055 | |||
| NM_178801.5 | NP_848916.2 | 15 | 2264 | |||
| GenBank mRNA | AK031517.1 | - | 16 | 2301 | ||
| AK038472.1 | - | 15 | 2160 | |||
| BC094382.1 | - | 15 | 2180 | 30605060 | ||
| BC137621.1 | - | 15 | 2147 | 8860640 | ||
| BC144788.1 | - | 13 | 1988 | |||

Tmem241
Genomic Map