siRNA Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178874.2 | NP_849205.1 | 5 | 3225 | ||
| GenBank mRNA | AK004359.1 | - | 1 | 977 | ||
| AK145948.1 | - | 3 | 3824 | |||
| AK157378.1 | - | 5 | 3338 | |||
| AK158436.1 | - | 4 | 2589 | |||
| AK165309.1 | - | 4 | 2586 | |||
| AK171635.1 | - | 4 | 2588 | |||
| AK172313.1 | - | 4 | 2586 | |||
| AK172356.1 | - | 4 | 2586 | |||
| AK172382.1 | - | 3 | 3822 | |||
| AK172541.1 | - | 4 | 2586 | |||
| AK172720.1 | - | 3 | 3824 | |||
| AK172957.1 | - | 5 | 3080 | |||
| BC052035.1 | - | 5 | 3225 | 5695005 | ||

Tmcc2
Genomic Map