siRNA Details
| Gene Symbol: Gorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178883.5 | NP_849214.2 | 5 | 1138 | ||
| GenBank mRNA | AK039170.1 | - | 5 | 1149 | ||
| AK043953.1 | - | 5 | 1139 | |||
| AK165909.1 | - | 5 | 1108 | |||
| AK169151.1 | - | 5 | 1131 | |||
| AK170455.1 | - | 6 | 1297 | |||
| BC013244.1 | - | 2 | 521 | |||
| BC013262.1 | - | 2 | 521 | |||
| BC119132.1 | - | 5 | 1121 | 8733881 | ||
| BC119134.1 | - | 5 | 1121 | 8733883 | ||

Gorab
Genomic Map