siRNA Details
| Gene Symbol: AI182371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243102.1 | NP_001230031.1 | 6 | 673 | ||
| NM_001243103.1 | NP_001230032.1 | 6 | 670 | |||
| NM_178885.4 | NP_849216.2 | 6 | 670 | |||
| GenBank mRNA | AK050126.1 | - | 6 | 674 | ||
| AK050413.1 | - | 6 | 645 | |||
| BC019541.1 | - | 6 | 628 | |||
| BC037995.1 | - | 6 | 650 | |||
| BC094504.1 | - | 6 | 658 | 5100182 | ||
| BC115979.1 | - | 6 | 645 | 40104352 | ||
| BC115986.1 | - | 6 | 645 | 40104349 | ||

AI182371
Genomic Map