siRNA Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178892.5 | NP_849223.2 | 6 | 1979 | ||
| GenBank mRNA | AK028529.1 | - | 6 | 1980 | ||
| AK032453.1 | - | 6 | 1963 | |||
| AK162094.1 | - | 6 | 1985 | |||
| AK163347.1 | - | 6 | 2019 | |||
| AK186495.1 | - | 2 | 340 | |||
| BC068173.1 | - | 6 | 1953 | 30616032 | ||

Tiparp
Genomic Map