siRNA Details
| Gene Symbol: AA792892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053948.1 | - | 1494 | |||
| AK087752.1 | - | 1555 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AA792892| Gene Symbol: AA792892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053948.1 | - | 1494 | |||
| AK087752.1 | - | 1555 | ||||