siRNA Details
Gene Symbol: Sprr2j-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019074.1 | - | 2 | 352 | ||
AY158994.1 | - | 2 | 355 | |||
BC125542.1 | - | 2 | 326 | |||
BC125544.1 | - | 2 | 326 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sprr2j-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019074.1 | - | 2 | 352 | ||
AY158994.1 | - | 2 | 355 | |||
BC125542.1 | - | 2 | 326 | |||
BC125544.1 | - | 2 | 326 |