siRNA Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159565.1 | NP_001153037.1 | 2 | 164 | ||
| NM_016890.4 | NP_058586.3 | 2 | 164 | |||
| GenBank mRNA | AF098508.1 | - | 2 | 143 | ||
| AK002720.1 | - | 2 | 155 | |||
| AK003255.1 | - | 2 | 139 | |||
| AK011840.1 | - | 2 | 161 | |||
| AK159344.1 | - | 2 | 162 | |||
| AV493721.1 | - | 2 | 150 | |||
| BC061120.1 | - | 2 | 145 | 6774742 | ||
| BY141790.1 | - | 2 | 171 | |||

Dctn3
Genomic Map