siRNA Details
| Gene Symbol: Ift20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018854.4 | NP_061342.1 | 3 | 485 | ||
| GenBank mRNA | AK002397.1 | - | 3 | 486 | ||
| AK139942.1 | - | 3 | 522 | |||
| AK213963.1 | - | 2 | 355 | |||
| AY082613.1 | - | 2 | 205 | |||
| BC002040.1 | - | 3 | 476 | 3590790 | ||
| U38981.1 | - | 318 | ||||

Ift20
Genomic Map