siRNA Details
| Gene Symbol: Ccl27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048179.1 | NP_001041644.1 | 6 | 491 | ||
| NM_011336.1 | NP_035466.1 | 3 | 325 | |||
| GenBank mRNA | AB013398.1 | - | 3 | 325 | ||
| AF082392.1 | - | 3 | 303 | |||
| AF099931.1 | - | 3 | 313 | |||
| AK005398.1 | - | 4 | 465 | |||
| AK146066.1 | - | 3 | 1829 | |||
| AY744154.1 | - | 1 | 633 | |||
| AY744155.1 | - | 2 | 444 | |||
| BC107226.2 | - | 3 | 325 | 40051915 | ||
| CA451425.1 | - | 6 | 491 | |||

Ccl27a
Genomic Map