siRNA Details
| Gene Symbol: Dysf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077694.2 | NP_001071162.1 | 3 | 491 | ||
| NM_021469.3 | NP_067444.2 | 3 | 309 | |||
| GenBank mRNA | AF188290.2 | - | 3 | 177 | ||
| AK131144.1 | - | 3 | 306 | |||
| BQ831003.1 | - | 365 | ||||
| CN716462.1 | - | 3 | 309 | |||
| EF041824.1 | - | 3 | 454 | |||

Dysf
Genomic Map