siRNA Details
Gene Symbol: Kcng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025734.2 | NP_080010.2 | 2 | 606 | ||
GenBank mRNA | AF450109.1 | - | 2 | 272 | ||
BC043936.1 | - | 2 | 606 | 4504272 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025734.2 | NP_080010.2 | 2 | 606 | ||
GenBank mRNA | AF450109.1 | - | 2 | 272 | ||
BC043936.1 | - | 2 | 606 | 4504272 |