siRNA Details
| Gene Symbol: Golm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035122.2 | NP_001030294.1 | 3 | 268 | ||
| NM_027307.4 | NP_081583.3 | 4 | 404 | |||
| GenBank mRNA | AK009072.1 | - | 2 | 75 | ||
| AK133865.1 | - | 3 | 270 | |||
| AK167646.1 | - | 3 | 245 | |||
| BC011152.1 | - | 3 | 229 | 4188434 | ||
| BC098486.1 | - | 4 | 400 | 30024953 | ||
| CD552063.1 | - | 4 | 365 | |||
| CK391733.1 | - | 3 | 268 | |||

Golm1
Genomic Map