siRNA Details
| Gene Symbol: Tex19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028602.2 | NP_082878.2 | 3 | 368 | ||
| GenBank mRNA | AF285590.1 | - | 3 | 368 | ||
| AK010735.1 | - | 3 | 355 | |||
| BC053492.1 | - | 3 | 334 | 30024269 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex19.1| Gene Symbol: Tex19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028602.2 | NP_082878.2 | 3 | 368 | ||
| GenBank mRNA | AF285590.1 | - | 3 | 368 | ||
| AK010735.1 | - | 3 | 355 | |||
| BC053492.1 | - | 3 | 334 | 30024269 | ||