siRNA Details
| Gene Symbol: Magea7-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC107020.2 | - | 1 | 135 | ||
| BC127031.1 | - | 1 | 136 | |||
| BC127032.1 | - | 1 | 135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magea7-ps| Gene Symbol: Magea7-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC107020.2 | - | 1 | 135 | ||
| BC127031.1 | - | 1 | 136 | |||
| BC127032.1 | - | 1 | 135 | |||