siRNA Details
| Gene Symbol: Ndufa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023312.2 | NP_075801.1 | 5 | 1062 | ||
| GenBank mRNA | AF286698.1 | - | 4 | 297 | ||
| AK002999.1 | - | 4 | 301 | |||
| AK003012.1 | - | 4 | 315 | |||
| AK010517.1 | - | 4 | 301 | |||
| AK012422.1 | - | 4 | 301 | |||
| BC034899.1 | - | 5 | 1062 | 1447015 | ||
| BC037149.1 | - | 4 | 305 | 5149812 | ||
| BC055435.1 | - | 4 | 311 | 4219489 | ||

Ndufa13
Genomic Map