siRNA Details
Gene Symbol: Rmnd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025346.1 | NP_079622.1 | 4, 5 | 788 | ||
GenBank mRNA | AK005344.1 | - | 1, 2 | 168 | ||
AK075599.1 | - | 4, 5 | 788 | |||
BC016075.1 | - | 4, 5 | 642 | 4485143 |
