siRNA Details
| Gene Symbol: Pink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026880.2 | NP_081156.2 | 1, 2 | 472 | ||
| GenBank mRNA | AB053476.1 | - | 1, 2 | 404 | ||
| AF316872.1 | - | 1, 2 | 450 | |||
| AK050322.1 | - | 1, 2 | 452 | |||
| AK155474.1 | - | 1, 2 | 458 | |||
| AK165480.1 | - | 1, 2 | 451 | |||
| AK187592.1 | - | 1, 2 | 33 | |||
| AK215426.1 | - | 1, 2 | 33 | |||
| AK220318.1 | - | 1, 2 | 426 | |||
| BC018561.1 | - | 1, 2 | 298 | |||
| BC018594.1 | - | 1, 2 | 435 | |||
| BC067066.1 | - | 1, 2 | 440 | 30542123 | ||
| BC069968.1 | - | 2, 3 | 330 | |||

Pink1
Genomic Map