siRNA Details
| Gene Symbol: Ndufb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023172.3 | NP_075661.1 | 2, 3 | 330 | ||
| GenBank mRNA | AK002254.1 | - | 2, 3 | 331 | ||
| AK003540.1 | - | 2, 3 | 331 | |||
| AK004522.1 | - | 2, 3 | 331 | |||
| BC031539.1 | - | 2, 3 | 312 | 3979845 | ||

Ndufb9
Genomic Map