siRNA Details
| Gene Symbol: Ubap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026872.1 | NP_081148.1 | 11 | 955 | ||
| GenBank mRNA | AF276965.1 | - | 10 | 859 | ||
| AK204028.1 | - | 3 | 106 | |||
| AK214539.1 | - | 1 | 46 | |||
| BC007179.1 | - | 11 | 955 | 3496608 | ||

Ubap2
Genomic Map