siRNA Details
| Gene Symbol: Ak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027592.3 | NP_081868.1 | 5 | 542 | ||
| GenBank mRNA | AK012905.1 | - | 5 | 492 | ||
| AK014903.1 | - | 4 | 464 | |||
| BC019453.1 | - | 5 | 493 | 4024616 | ||
| BG084768.2 | - | 3 | 329 | |||

Ak6
Genomic Map