siRNA Details
| Gene Symbol: Bcor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168321.1 | NP_001161793.1 | 2, 3 | 200 | ||
| NM_029510.3 | NP_083786.2 | 2, 3 | 869 | |||
| NM_175044.3 | NP_778209.2 | 2, 3 | 869 | |||
| NM_175045.3 | NP_778210.2 | 2, 3 | 869 | |||
| NM_175046.3 | NP_778211.2 | 2, 3 | 869 | |||
| GenBank mRNA | AK053309.1 | - | 2, 3 | 870 | ||
| AK086385.2 | - | 2, 3 | 376 | |||
| AK129398.1 | - | 2, 3 | 200 | |||
| AY161170.1 | - | 2, 3 | 184 | |||
| AY161171.1 | - | 2, 3 | 184 | |||
| AY161172.1 | - | 2, 3 | 184 | |||
| AY161173.1 | - | 2, 3 | 184 | |||
| BC058656.1 | - | 2, 3 | 370 | 6412868 | ||

Bcor
Genomic Map