siRNA Details
| Gene Symbol: Mospd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254762.1 | NP_001241691.1 | 2 | 490 | ||
| NM_030037.2 | NP_084313.1 | 3 | 643 | |||
| GenBank mRNA | AK088731.1 | - | 2 | 481 | ||
| BC003880.1 | - | 2 | 500 | 3496491 | ||
| BC034167.1 | - | 3 | 628 | |||
| BC039620.1 | - | 3 | 624 | 5051528 | ||
| BE861793.1 | - | 2 | 117 | |||
| BQ445267.1 | - | 1 | 56 | |||
| CK617131.1 | - | 2 | 441 | |||

Mospd3
Genomic Map