siRNA Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030565.6 | NP_085042.2 | 10 | 2099 | ||
| GenBank mRNA | AK149780.1 | - | 11 | 2179 | ||
| AK154810.1 | - | 10 | 2095 | |||
| AK220286.1 | - | 10 | 1615 | |||
| AY778962.1 | - | 10 | 2080 | |||
| BC004044.1 | - | 7 | 708 | |||
| BC025814.1 | - | 10 | 2050 | |||
| BC025826.1 | - | 10 | 2080 | |||
| CT010363.1 | - | 6 | 554 | |||

Fam20c
Genomic Map