siRNA Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133925.2 | NP_598686.2 | 8 | 925 | ||
| GenBank mRNA | AK156773.1 | - | 8 | 862 | ||
| AK161670.1 | - | 8 | 891 | |||
| AK167050.1 | - | 8 | 927 | |||
| BC018373.1 | - | 4 | 299 | 3708613 | ||
| BC024602.1 | - | 8 | 806 | |||
| BC027123.1 | - | 1 | 231 | 5356457 | ||
| BC040811.1 | - | 8 | 870 | 4015830 | ||

Rbm28
Genomic Map