siRNA Details
| Gene Symbol: Otop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172709.3 | NP_766297.2 | 3 | 593 | ||
| GenBank mRNA | AF548337.1 | - | 3 | 593 | ||
| AK040792.1 | - | 3 | 580 | |||
| AK041023.1 | - | 3 | 367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otop1| Gene Symbol: Otop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172709.3 | NP_766297.2 | 3 | 593 | ||
| GenBank mRNA | AF548337.1 | - | 3 | 593 | ||
| AK040792.1 | - | 3 | 580 | |||
| AK041023.1 | - | 3 | 367 | |||