siRNA Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029012.2 | NP_083288.2 | 10, 11 | 1160 | ||
| GenBank mRNA | AK014709.1 | - | 10, 11 | 1498 | ||
| BC023131.1 | - | 6, 7 | 775 | |||
| BC078437.1 | - | 1, 2 | 1207 | |||
| BC086646.1 | - | 10, 11 | 1525 | 6817386 | ||

Sppl3
Genomic Map