siRNA Details
| Gene Symbol: Mrpl47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029017.2 | NP_083293.1 | 2 | 212 | ||
| GenBank mRNA | BC029173.1 | - | 192 | |||
| BC038065.1 | - | 2 | 183 | |||
| BC087953.1 | - | 2 | 206 | 30119240 | ||
| BY013327.1 | - | 2 | 212 | |||
| CX122942.1 | - | 2 | 191 | |||

Mrpl47
Genomic Map