siRNA Details
| Gene Symbol: Chmp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029362.3 | NP_083638.1 | 1 | 284 | ||
| GenBank mRNA | AK008205.1 | - | 1 | 249 | ||
| AK156473.1 | - | 1 | 244 | |||
| AK159193.1 | - | 1 | 246 | |||
| BC011429.1 | - | 1 | 261 | 3485793 | ||
| BC059279.1 | - | 1 | 236 | 4019739 | ||
| CK628208.1 | - | 1 | 284 | |||

Chmp4b
Genomic Map