siRNA Details
| Gene Symbol: 3110002H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029623.2 | NP_083899.1 | 2 | 227 | ||
| GenBank mRNA | AK010309.1 | - | 2 | 221 | ||
| AK144623.1 | - | 2 | 228 | |||
| AK154240.1 | - | 2 | 222 | |||
| AK159691.1 | - | 2 | 222 | |||
| BC012285.1 | - | 2 | 205 | |||
| BC021846.1 | - | 2 | 210 | 5027035 | ||

3110002H16Rik
Genomic Map