siRNA Details
| Gene Symbol: Cwc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290740.1 | NP_001277669.1 | 4 | 493 | ||
| NM_030560.5 | NP_085037.2 | 5 | 551 | |||
| NM_172667.2 | NP_766255.1 | 4 | 490 | |||
| GenBank mRNA | AK045589.1 | - | 4 | 322 | ||
| AK051237.1 | - | 4 | 318 | |||
| AK077961.1 | - | 5 | 379 | |||
| AK088752.1 | - | 4 | 321 | |||
| AK089988.1 | - | 326 | ||||
| AK129404.1 | - | 304 | ||||
| AK132001.1 | - | 4 | 434 | |||
| AK143352.1 | - | 4 | 323 | |||
| AK149528.1 | - | 4 | 317 | |||
| AK166487.1 | - | 4 | 326 | |||
| BC003993.1 | - | 4 | 303 | 3492146 | ||
| BU613482.1 | - | 4 | 490 | |||

Cwc22
Genomic Map