siRNA Details
| Gene Symbol: Il17f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145856.2 | NP_665855.2 | 3 | 412 | ||
| GenBank mRNA | AB116259.1 | - | 3 | 341 | ||
| AF458064.1 | - | 2 | 317 | |||
| AK157101.1 | - | 1 | 81 | |||
| AY380822.1 | - | 3 | 412 | |||
| BC145977.1 | - | 2 | 357 | |||

Il17f
Genomic Map