siRNA Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153502.3 | NP_705722.2 | 2, 3 | 196 | ||
| GenBank mRNA | AF492400.1 | - | 2, 3 | 196 | ||
| BC022973.1 | - | 355 | ||||
| BC023221.1 | - | 2, 3 | 173 | |||
| BC132370.1 | - | 2, 3 | 206 | 40130647 | ||
| BC138797.1 | - | 2, 3 | 206 | 8861819 | ||
| BC145421.1 | - | 2, 3 | 206 | 9053979 | ||

Ankrd23
Genomic Map