siRNA Details
| Gene Symbol: Acyp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029344.3 | NP_083620.1 | 3 | 131 | ||
| GenBank mRNA | AK009134.1 | - | 3 | 131 | ||
| AK046238.1 | - | 3 | 140 | |||
| BC027642.1 | - | 3 | 119 | 1245384 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acyp2| Gene Symbol: Acyp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029344.3 | NP_083620.1 | 3 | 131 | ||
| GenBank mRNA | AK009134.1 | - | 3 | 131 | ||
| AK046238.1 | - | 3 | 140 | |||
| BC027642.1 | - | 3 | 119 | 1245384 | ||